siRNA Details
| Gene Symbol: Tmem194b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142647.1 | NP_001136119.1 | 9 | 1771 | ||
| GenBank mRNA | AK030361.1 | - | 8 | 1626 | ||
| AK036693.1 | - | 9 | 1685 | |||
| AK086801.1 | - | 5 | 1168 | |||
| AK136177.1 | - | 5 | 1259 | |||
| AK170577.1 | - | 4 | 3075 | |||
| BC094370.1 | - | 4 | 2715 | |||

Tmem194b
Genomic Map