siRNA Details
| Gene Symbol: D2Wsu81e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172660.4 | NP_766248.3 | 12 | 1688 | ||
| GenBank mRNA | AK036945.1 | - | 10 | 1982 | ||
| AK044983.1 | - | 10 | 2282 | |||
| AK046148.1 | - | 11 | 1883 | |||
| AK147162.1 | - | 12 | 1678 | |||
| AK218621.1 | - | 1 | 81 | |||
| BC078441.1 | - | 9 | 2270 | 5686308 | ||
| BY462424.1 | - | 1 | 280 | |||

D2Wsu81e
Genomic Map