siRNA Details
| Gene Symbol: Gtdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172662.3 | NP_766250.2 | 9, 10 | 1357 | ||
| GenBank mRNA | AK054272.1 | - | 9, 10 | 1358 | ||
| AK162289.1 | - | 1, 2 | 144 | |||
| BC052069.1 | - | 6, 7 | 922 | 5715781 | ||

Gtdc1
Genomic Map