siRNA Details
| Gene Symbol: Epc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172663.4 | NP_766251.3 | 2 | 616 | ||
| GenBank mRNA | AK031025.1 | - | 2 | 589 | ||
| AK047947.1 | - | 2 | 284 | |||
| BB620197.1 | - | 2 | 579 | |||
| BC048785.1 | - | 1 | 55 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Epc2| Gene Symbol: Epc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172663.4 | NP_766251.3 | 2 | 616 | ||
| GenBank mRNA | AK031025.1 | - | 2 | 589 | ||
| AK047947.1 | - | 2 | 284 | |||
| BB620197.1 | - | 2 | 579 | |||
| BC048785.1 | - | 1 | 55 | |||