siRNA Details
| Gene Symbol: Ambra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080754.1 | NP_001074223.1 | 8 | 1476 | ||
| NM_172669.3 | NP_766257.3 | 7 | 1749 | |||
| GenBank mRNA | AK037516.1 | - | 7 | 1706 | ||
| AK083596.1 | - | 7 | 1706 | |||
| AK084557.1 | - | 7 | 1731 | |||
| AK129433.1 | - | 3 | 1013 | |||
| AK148887.1 | - | 8 | 1468 | |||
| AK155513.1 | - | 7 | 1712 | |||
| AK169280.1 | - | 8 | 1429 | |||
| BC032293.1 | - | 8 | 1399 | |||
| BC054082.1 | - | 7 | 1681 | |||

Ambra1
Genomic Map