siRNA Details
| Gene Symbol: Frmd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172673.3 | NP_766261.2 | 7, 8 | 864 | ||
| GenBank mRNA | AK030883.1 | - | 7, 8 | 620 | ||
| AK044270.1 | - | 7, 8 | 615 | |||
| AK048865.1 | - | 7, 8 | 865 | |||
| AK054484.1 | - | 7, 8 | 821 | |||
| BC038623.1 | - | 7, 8 | 601 | |||
| BC057549.1 | - | 4, 5 | 393 | 6416914 | ||
| BC062889.1 | - | 7, 8 | 821 | 5698263 | ||

Frmd5
Genomic Map