siRNA Details
| Gene Symbol: Frmd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172673.3 | NP_766261.2 | 13 | 1286 | ||
| GenBank mRNA | AK030883.1 | - | 13 | 1042 | ||
| AK044270.1 | - | 13 | 1037 | |||
| BC038623.1 | - | 13 | 1023 | |||
| BC057549.1 | - | 10 | 815 | 6416914 | ||
| BC062889.1 | - | 13 | 1243 | 5698263 | ||

Frmd5
Genomic Map