siRNA Details
| Gene Symbol: Slc25a24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172685.3 | NP_766273.1 | 3 | 439 | ||
| GenBank mRNA | AK032806.1 | - | 3 | 440 | ||
| AK146034.1 | - | 3 | 438 | |||
| AK167778.1 | - | 3 | 430 | |||
| AK168090.1 | - | 3 | 430 | |||
| AK170598.1 | - | 3 | 431 | |||
| AK187917.1 | - | 2 | 208 | |||
| BC055369.1 | - | 3 | 428 | 30055307 | ||

Slc25a24
Genomic Map