siRNA Details
| Gene Symbol: N28178 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172690.2 | NP_766278.1 | 4 | 528 | ||
| GenBank mRNA | AK038528.1 | - | 3 | 382 | ||
| AK046283.1 | - | 4 | 519 | |||
| AK122428.1 | - | 2 | 451 | |||
| BC057092.1 | - | 4 | 528 | 5703966 | ||

N28178
Genomic Map