siRNA Details
| Gene Symbol: Fam214b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253353.1 | NP_001240282.1 | 3 | 849 | ||
| NM_001253354.1 | NP_001240283.1 | 4 | 626 | |||
| NM_172691.3 | NP_766279.1 | 3 | 630 | |||
| GenBank mRNA | AK045811.1 | - | 3 | 614 | ||
| AK149610.1 | - | 3 | 707 | |||
| AK154748.1 | - | 2 | 965 | |||
| AK161380.1 | - | 3 | 557 | |||
| BC044832.1 | - | 2 | 790 | |||
| BC046771.1 | - | 4 | 616 | 5703628 | ||
| BC054736.1 | - | 3 | 630 | |||
| BC071191.1 | - | 1 | 318 | 6467750 | ||

Fam214b
Genomic Map