siRNA Details
| Gene Symbol: Fam214b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253353.1 | NP_001240282.1 | 4, 5 | 1514 | ||
| NM_001253354.1 | NP_001240283.1 | 5, 6 | 1291 | |||
| NM_172691.3 | NP_766279.1 | 4, 5 | 1295 | |||
| GenBank mRNA | AK045811.1 | - | 4, 5 | 1279 | ||
| AK149610.1 | - | 4, 5 | 1372 | |||
| AK154748.1 | - | 3, 4 | 1630 | |||
| AK161380.1 | - | 4, 5 | 1222 | |||
| AK173197.1 | - | 1, 2 | 40 | |||
| BC046771.1 | - | 5, 6 | 1281 | 5703628 | ||
| BC071191.1 | - | 2, 3 | 983 | 6467750 | ||

Fam214b
Genomic Map