siRNA Details
| Gene Symbol: Dnajc11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172704.3 | NP_766292.2 | 2 | 209 | ||
| GenBank mRNA | AK053156.1 | - | 2 | 191 | ||
| AK089983.1 | - | 2 | 201 | |||
| AK156793.1 | - | 2 | 191 | |||
| AK171781.1 | - | 2 | 190 | |||
| AK172542.1 | - | 2 | 190 | |||
| BC075619.1 | - | 2 | 185 | |||
| BC079852.1 | - | 2 | 179 | |||
| BC085257.1 | - | 2 | 183 | 5414963 | ||
| BY784439.1 | - | 2 | 220 | |||

Dnajc11
Genomic Map