siRNA Details
| Gene Symbol: Phf13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172705.2 | NP_766293.2 | 4 | 1330 | ||
| GenBank mRNA | AK077052.1 | - | 2 | 844 | ||
| AK132066.1 | - | 4 | 1335 | |||
| AY069976.2 | - | 4 | 1255 | |||
| BC020286.1 | - | 2 | 379 | |||
| BC029632.1 | - | 4 | 1330 | 3500487 | ||
| BC059282.1 | - | 4 | 1330 | 3500487 | ||

Phf13
Genomic Map