siRNA Details
| Gene Symbol: Sel1l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172710.3 | NP_766298.2 | 2 | 757 | ||
| GenBank mRNA | AK030487.1 | - | 2 | 622 | ||
| AK031770.1 | - | 3 | 621 | |||
| AK037220.1 | - | 2 | 621 | |||
| AK122362.1 | - | 2 | 757 | |||
| BC094607.1 | - | 1 | 302 | 6837048 | ||
| BC117915.1 | - | 1 | 258 | 40097358 | ||
| BC117916.1 | - | 1 | 260 | 40097359 | ||

Sel1l3
Genomic Map