siRNA Details
| Gene Symbol: Clec9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205363.1 | NP_001192292.1 | 7 | 1582 | ||
| NM_001205364.1 | NP_001192293.1 | 6 | 1501 | |||
| NM_001205365.1 | NP_001192294.1 | 4 | 1233 | |||
| NM_172732.3 | NP_766320.2 | 6 | 1504 | |||
| GenBank mRNA | AK036399.1 | - | 7 | 1584 | ||

Clec9a
Genomic Map