siRNA Details
| Gene Symbol: Dera | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172733.1 | NP_766321.1 | 3 | 187 | ||
| GenBank mRNA | AK012558.1 | - | 3 | 223 | ||
| AK030961.1 | - | 3 | 219 | |||
| AK034347.1 | - | 189 | ||||
| AK075963.1 | - | 3 | 212 | |||
| AK083298.1 | - | 3 | 187 | |||
| AK145820.1 | - | 3 | 223 | |||
| AK150273.1 | - | 3 | 213 | |||
| AK150712.1 | - | 3 | 183 | |||
| AK152462.1 | - | 3 | 191 | |||
| AK168204.1 | - | 3 | 223 | |||
| AK169418.1 | - | 3 | 207 | |||
| BC016218.1 | - | 3 | 193 | 4484163 | ||

Dera
Genomic Map