siRNA Details
| Gene Symbol: Dera | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172733.1 | NP_766321.1 | 5, 6 | 547 | ||
| GenBank mRNA | AK012558.1 | - | 5, 6 | 583 | ||
| AK034347.1 | - | 549 | ||||
| AK075963.1 | - | 5, 6 | 572 | |||
| AK083298.1 | - | 5, 6 | 547 | |||
| AK145820.1 | - | 5, 6 | 583 | |||
| AK150273.1 | - | 5, 6 | 573 | |||
| AK150712.1 | - | 5, 6 | 543 | |||
| AK152462.1 | - | 5, 6 | 551 | |||
| AK168204.1 | - | 5, 6 | 583 | |||
| AK169418.1 | - | 5, 6 | 567 | |||
| AK179075.1 | - | 1, 2 | 52 | |||
| BC016218.1 | - | 5, 6 | 553 | 4484163 | ||

Dera
Genomic Map