siRNA Details
| Gene Symbol: Mtmr10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172742.2 | NP_766330.2 | 16 | 2023 | ||
| GenBank mRNA | AK028584.1 | - | 4 | 648 | ||
| BC025088.1 | - | 1 | 85 | |||
| BC041672.1 | - | 16 | 1995 | |||
| BC055074.1 | - | 14 | 1687 | 30052208 | ||
| BC058196.1 | - | 9 | 1125 | |||

Mtmr10
Genomic Map