siRNA Details
| Gene Symbol: Sprr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173070.1 | NP_775093.1 | 1 | 218 | ||
| GenBank mRNA | AY158996.1 | - | 1 | 218 | ||
| BC107019.1 | - | 1 | 218 | 40049167 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sprr4| Gene Symbol: Sprr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173070.1 | NP_775093.1 | 1 | 218 | ||
| GenBank mRNA | AY158996.1 | - | 1 | 218 | ||
| BC107019.1 | - | 1 | 218 | 40049167 | ||