siRNA Details
| Gene Symbol: Csnk1g1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173185.2 | NP_775277.2 | 10 | 1383 | ||
| GenBank mRNA | AK090262.1 | - | 10 | 1384 | ||
| BC064645.1 | - | 9 | 1399 | 5363779 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Csnk1g1| Gene Symbol: Csnk1g1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173185.2 | NP_775277.2 | 10 | 1383 | ||
| GenBank mRNA | AK090262.1 | - | 10 | 1384 | ||
| BC064645.1 | - | 9 | 1399 | 5363779 | ||