siRNA Details
| Gene Symbol: 2310035C23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029349.1 | NP_083625.1 | 1 | 555 | ||
| NM_173187.3 | NP_775279.2 | 1 | 555 | |||
| GenBank mRNA | AK009628.1 | - | 1 | 480 | ||
| AK035724.1 | - | 1 | 557 | |||
| AK129368.1 | - | 1 | 528 | |||
| AK148005.1 | - | 1 | 534 | |||
| BC051567.1 | - | 1 | 506 | |||
| BC117949.1 | - | 1 | 489 | 40098078 | ||
| BC117950.1 | - | 1 | 493 | 40098082 | ||

2310035C23Rik
Genomic Map