siRNA Details
| Gene Symbol: 2310035C23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029349.1 | NP_083625.1 | 2 | 794 | ||
| NM_173187.3 | NP_775279.2 | 2 | 794 | |||
| GenBank mRNA | AK035724.1 | - | 2 | 796 | ||
| AK040168.1 | - | 3 | 430 | |||
| AK129368.1 | - | 2 | 767 | |||
| AK148005.1 | - | 2 | 774 | |||
| BC117949.1 | - | 2 | 728 | 40098078 | ||
| BC117950.1 | - | 2 | 732 | 40098082 | ||

2310035C23Rik
Genomic Map