siRNA Details
| Gene Symbol: Proser1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173382.1 | NP_775558.2 | 13 | 3858 | ||
| GenBank mRNA | AK034174.1 | - | 13 | 4016 | ||
| AK038404.1 | - | 13 | 3858 | |||
| AK049354.1 | - | 3 | 1043 | |||
| BC051453.1 | - | 3 | 2626 | |||
| BC057544.1 | - | 5 | 2768 | |||
| BC079616.1 | - | 2628 | ||||
| BC086674.1 | - | 13 | 3415 | 30653602 | ||

Proser1
Genomic Map