siRNA Details
| Gene Symbol: Gpr171 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173398.3 | NP_775574.1 | 2 | 435 | ||
| GenBank mRNA | AK041381.1 | - | 2 | 407 | ||
| AK041582.1 | - | 2 | 431 | |||
| AK079674.1 | - | 2 | 436 | |||
| AK083323.1 | - | 2 | 432 | |||
| AK089287.1 | - | 2 | 388 | |||
| AK154725.1 | - | 2 | 433 | |||
| BC024054.1 | - | 3 | 361 | 5068166 | ||
More Information
Set Membership: |
|

Gpr171
Genomic Map