siRNA Details
| Gene Symbol: Haus6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173400.2 | NP_775576.2 | 17 | 3492 | ||
| GenBank mRNA | AK036635.1 | - | 8 | 2159 | ||
| AK169222.1 | - | 17 | 3495 | |||
| AK173206.1 | - | 16 | 4918 | |||
| BC044803.1 | - | 2 | 1253 | |||
| BC068237.1 | - | 17 | 3492 | 30550558 | ||

Haus6
Genomic Map