siRNA Details
| Gene Symbol: Fbxo44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161851.2 | NP_001155323.1 | 4 | 538 | ||
| NM_173401.5 | NP_775577.2 | 4 | 815 | |||
| GenBank mRNA | AK019407.1 | - | 4 | 570 | ||
| AK077452.1 | - | 4 | 601 | |||
| AK170958.1 | - | 4 | 498 | |||
| AK191511.1 | - | 3 | 411 | |||
| AK218805.1 | - | 3 | 344 | |||
| BC028884.1 | - | 4 | 607 | 4165174 | ||
| CX202860.1 | - | 3 | 239 | |||

Fbxo44
Genomic Map