siRNA Details
Gene Symbol: Jazf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168277.1 | NP_001161749.1 | 2 | 227 | ||
NM_173406.3 | NP_775582.2 | 2 | 226 | |||
GenBank mRNA | AK043767.1 | - | 2 | 176 | ||
AK050474.1 | - | 2 | 198 | |||
BC048577.1 | - | 2 | 227 | 6706223 | ||
BY278789.1 | - | 2 | 226 |
