siRNA Details
| Gene Symbol: Hmces | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173737.2 | NP_776098.1 | 4 | 412 | ||
| GenBank mRNA | AK010550.1 | - | 2 | 180 | ||
| AK018400.1 | - | 3 | 562 | |||
| BC024401.1 | - | 4 | 412 | 5026919 | ||
| BC064070.1 | - | 4 | 395 | 3371288 | ||

Hmces
Genomic Map