siRNA Details
Gene Symbol: Ilvbl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173751.4 | NP_776112.1 | 11, 12 | 1474 | ||
GenBank mRNA | AK087956.1 | - | 10, 11 | 1348 | ||
AK146075.1 | - | 10, 11 | 1402 | |||
AK169899.1 | - | 10, 11 | 1400 | |||
AK184668.1 | - | 2, 3 | 180 | |||
BC052054.1 | - | 10, 11 | 1808 | |||
BC056459.1 | - | 11, 12 | 1432 | 5718157 |
