siRNA Details
| Gene Symbol: Tapt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173764.3 | NP_776125.2 | 8 | 929 | ||
| GenBank mRNA | AK030002.1 | - | 3 | 171 | ||
| BC096038.1 | - | 5 | 408 | |||
| BC110670.1 | - | 7 | 650 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tapt1| Gene Symbol: Tapt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173764.3 | NP_776125.2 | 8 | 929 | ||
| GenBank mRNA | AK030002.1 | - | 3 | 171 | ||
| BC096038.1 | - | 5 | 408 | |||
| BC110670.1 | - | 7 | 650 | |||