siRNA Details
| Gene Symbol: Npr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173788.3 | NP_776149.1 | 1 | 660 | ||
| GenBank mRNA | AF331708.1 | - | 1 | 345 | ||
| BC042470.1 | - | 1 | 660 | 5137261 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Npr2| Gene Symbol: Npr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173788.3 | NP_776149.1 | 1 | 660 | ||
| GenBank mRNA | AF331708.1 | - | 1 | 345 | ||
| BC042470.1 | - | 1 | 660 | 5137261 | ||