siRNA Details
| Gene Symbol: Crybg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174848.3 | NP_777273.2 | 18, 19 | 3641 | ||
| GenBank mRNA | AK030282.1 | - | 9, 10 | 1085 | ||
| AK162506.1 | - | 6, 7 | 1476 | |||
| BC043118.1 | - | 18, 19 | 3641 | 6408859 | ||

Crybg3
Genomic Map