siRNA Details
| Gene Symbol: TSNAX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005999.2 | NP_005990.1 | 2 | 242 | ||
| GenBank mRNA | AK313068.1 | - | 2 | 180 | ||
| BC010376.1 | - | 2 | 200 | 4283594 | ||
| BC011797.1 | - | 2 | 217 | 4138517 | ||
| X95073.1 | - | 2 | 244 | |||
| Gene Symbol: TSNAX-DISC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028393.1 | 2 | 242 | |||
| NR_028394.1 | 2 | 242 | ||||
| NR_028395.1 | 2 | 242 | ||||
| NR_028396.1 | 2 | 242 | ||||
| NR_028397.1 | 2 | 242 | ||||
| NR_028398.1 | 2 | 242 | ||||
| NR_028399.1 | 2 | 242 | ||||
| NR_028400.1 | 2 | 242 | ||||

TSNAX
Genomic Map