siRNA Details
| Gene Symbol: Fam73a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162375.1 | NP_001155847.1 | 14 | 2392 | ||
| NM_174868.4 | NP_777357.2 | 16 | 2584 | |||
| GenBank mRNA | AK045347.1 | - | 14 | 2391 | ||
| AK048575.1 | - | 1 | 278 | |||
| AK083909.1 | - | 15 | 2415 | |||
| AK085017.1 | - | 16 | 2584 | |||
| AK143366.1 | - | 16 | 2584 | |||
| AK157801.1 | - | 1 | 71 | |||
| AK166051.1 | - | 16 | 2537 | |||
| BC098205.1 | - | 16 | 2567 | 6409928 | ||

Fam73a
Genomic Map