siRNA Details
| Gene Symbol: Pou6f2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175006.2 | NP_778171.2 | 2 | 587 | ||
| GenBank mRNA | AK083772.1 | - | 2 | 587 | ||
| BC094915.1 | - | 2 | 602 | 6435959 | ||
| BC138151.1 | - | 2 | 506 | 8861170 | ||
| BG807013.1 | - | 2 | 237 | |||

Pou6f2
Genomic Map