siRNA Details
| Gene Symbol: Pou6f2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175006.2 | NP_778171.2 | 4, 5 | 1154 | ||
| GenBank mRNA | AK042530.1 | - | 1, 2 | 132 | ||
| AK083772.1 | - | 4, 5 | 1154 | |||
| BC094915.1 | - | 4, 5 | 1169 | 6435959 | ||
| BC138151.1 | - | 4, 5 | 1076 | 8861170 | ||

Pou6f2
Genomic Map