siRNA Details
| Gene Symbol: Eny2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175009.3 | NP_778174.1 | 3 | 263 | ||
| GenBank mRNA | AF173297.1 | - | 3 | 283 | ||
| AK007877.1 | - | 3 | 214 | |||
| AK016562.1 | - | 3 | 1648 | |||
| AK032957.1 | - | 3 | 263 | |||
| AK160628.1 | - | 3 | 231 | |||
| AK162511.1 | - | 3 | 260 | |||
| BC048361.1 | - | 3 | 269 | 4012930 | ||

Eny2
Genomic Map