siRNA Details
| Gene Symbol: 1110051M20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175123.4 | NP_780332.1 | 2 | 140 | ||
| GenBank mRNA | AK018339.1 | - | 2 | 174 | ||
| AK045977.1 | - | 2 | 141 | |||
| AK164320.1 | - | 2 | 138 | |||
| BC032255.1 | - | 2 | 160 | |||
| BC085239.1 | - | 2 | 139 | 6440075 | ||
| BC089006.1 | - | 2 | 120 | 6843427 | ||

1110051M20Rik
Genomic Map