siRNA Details
| Gene Symbol: 1110051M20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175123.4 | NP_780332.1 | 3 | 194 | ||
| GenBank mRNA | AK018339.1 | - | 3 | 228 | ||
| AK045977.1 | - | 3 | 195 | |||
| AK164320.1 | - | 3 | 192 | |||
| BC032255.1 | - | 3 | 214 | |||
| BC049648.1 | - | 2 | 129 | |||
| BC085239.1 | - | 3 | 193 | 6440075 | ||
| BC089006.1 | - | 3 | 174 | 6843427 | ||

1110051M20Rik
Genomic Map