siRNA Details
| Gene Symbol: Zmat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175446.3 | NP_780655.2 | 7 | 1102 | ||
| GenBank mRNA | AK039100.1 | - | 3 | 2208 | ||
| AK046974.1 | - | 2 | 1234 | |||
| AK133258.1 | - | 1099 | ||||
| AK220419.1 | - | 6 | 743 | |||
| BC013473.1 | - | 4 | 455 | |||
| BC067022.1 | - | 7 | 1078 | |||
| BC127247.1 | - | 2 | 671 | |||
| BC128569.1 | - | 2 | 671 | |||

Zmat1
Genomic Map