siRNA Details
| Gene Symbol: Glis3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY220846.1 | - | 4, 5 | 1600 | ||
| BC066143.1 | - | 5, 6 | 1935 | |||
| BC129906.1 | - | 4, 5 | 1578 | |||
| BC129907.1 | - | 4, 5 | 1578 | |||
| DQ871027.1 | - | 6, 7 | 2555 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Glis3| Gene Symbol: Glis3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY220846.1 | - | 4, 5 | 1600 | ||
| BC066143.1 | - | 5, 6 | 1935 | |||
| BC129906.1 | - | 4, 5 | 1578 | |||
| BC129907.1 | - | 4, 5 | 1578 | |||
| DQ871027.1 | - | 6, 7 | 2555 | |||