siRNA Details
| Gene Symbol: Fam78b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160261.1 | NP_001153733.1 | 2 | 651 | ||
| NM_001160262.1 | NP_001153734.1 | 2 | 651 | |||
| NM_175461.4 | NP_780670.2 | 2 | 651 | |||
| GenBank mRNA | AK033840.1 | - | 1 | 149 | ||
| AK047703.1 | - | 2 | 489 | |||
| AK144212.1 | - | 2 | 605 | |||
| BC066094.1 | - | 2 | 479 | |||
| BC132161.1 | - | 3 | 574 | 40130438 | ||
| BC132525.1 | - | 2 | 636 | 40130802 | ||

Fam78b
Genomic Map