siRNA Details
| Gene Symbol: Kcnt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK033953.1 | - | 5 | 407 | ||
| AK086119.1 | - | 4 | 380 | |||
| AK147625.1 | - | 3 | 2357 | |||
| BB662543.1 | - | 4 | 375 | |||
| BC141190.1 | - | 4 | 453 | 9055802 | ||
| BC171963.1 | - | 4 | 453 | 9054249 | ||
| FJ628361.1 | - | 5 | 451 | |||

Kcnt1
Genomic Map