siRNA Details
| Gene Symbol: ST8SIA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006011.3 | NP_006002.1 | 2 | 272 | ||
| XM_017022642.1 | XP_016878131.1 | 2 | 435 | |||
| GenBank mRNA | AK313924.1 | - | 2 | 220 | ||
| AW163487.1 | - | 272 | ||||
| BC069584.1 | - | 2 | 141 | |||
| BC096202.3 | - | 2 | 141 | 40004644 | ||
| BC096203.3 | - | 2 | 141 | 40004645 | ||
| BC096204.3 | - | 2 | 141 | 40004646 | ||
| U33551.1 | - | 2 | 117 | |||
| U82762.1 | - | 2 | 142 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_009181, X83562, BC096546.

ST8SIA2
Genomic Map