siRNA Details
Gene Symbol: Cyb5rl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175471.2 | NP_780680.1 | 2 | 460 | ||
GenBank mRNA | AK042316.1 | - | 3 | 2074 | ||
AK053031.1 | - | 5 | 864 | |||
AK220154.1 | - | 3 | 362 | |||
AY437136.1 | - | 290 | ||||
BC004750.1 | - | 2 | 299 | |||
BC043687.1 | - | 2 | 460 | 5136046 |
