siRNA Details
Gene Symbol: Cyb5rl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175471.2 | NP_780680.1 | 6 | 1039 | ||
GenBank mRNA | AK053031.1 | - | 9 | 1444 | ||
AK220154.1 | - | 8 | 1367 | |||
BC004750.1 | - | 5 | 773 | |||
BC043687.1 | - | 6 | 1039 | 5136046 |
