siRNA Details
| Gene Symbol: Fam109a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175474.3 | NP_780683.1 | 4 | 2314 | ||
| GenBank mRNA | AK033618.1 | - | 4 | 2315 | ||
| AK039192.1 | - | 3 | 2262 | |||
| AK081460.1 | - | 1 | 1435 | |||
| AK186609.1 | - | 1 | 265 | |||
| BC095929.1 | - | 739 | ||||

Fam109a
Genomic Map