siRNA Details
| Gene Symbol: Zfp574 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168506.1 | NP_001161978.1 | 2 | 1809 | ||
| NM_175477.4 | NP_780686.1 | 2 | 1864 | |||
| GenBank mRNA | AK042074.1 | - | 2 | 1827 | ||
| AK053744.1 | - | 2 | 1750 | |||
| AK143997.1 | - | 2 | 1810 | |||
| AK170243.1 | - | 2 | 1715 | |||
| BC021833.2 | - | 2 | 1809 | |||
| BC054064.1 | - | 2 | 1765 | |||
| BC059044.1 | - | 2 | 1735 | 6816316 | ||
| BC094354.1 | - | 2 | 1789 | |||

Zfp574
Genomic Map