siRNA Details
| Gene Symbol: Cbln4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175631.3 | NP_783439.1 | 3 | 2400 | ||
| GenBank mRNA | AK032406.1 | - | 3 | 1904 | ||
| AK032621.1 | - | 3 | 2403 | |||
| BC094540.1 | - | 3 | 2303 | 5708067 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cbln4| Gene Symbol: Cbln4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175631.3 | NP_783439.1 | 3 | 2400 | ||
| GenBank mRNA | AK032406.1 | - | 3 | 1904 | ||
| AK032621.1 | - | 3 | 2403 | |||
| BC094540.1 | - | 3 | 2303 | 5708067 | ||