siRNA Details
| Gene Symbol: Vwa3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK038906.1 | - | 4 | 623 | ||
| AK050189.1 | - | 4 | 489 | |||
| BC049712.1 | - | 4 | 634 | |||
| BC147491.1 | - | 4 | 483 | 9087438 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vwa3b| Gene Symbol: Vwa3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK038906.1 | - | 4 | 623 | ||
| AK050189.1 | - | 4 | 489 | |||
| BC049712.1 | - | 4 | 634 | |||
| BC147491.1 | - | 4 | 483 | 9087438 | ||